Learn More
Description
The MOPC-173 antibody has unknown specificity. The transplantable plasmacytoma MOPC-173 was induced by intraperitoneal injection of mineral oils into BALB/c mice. In the absence of antigen-specific binding, this immunoglobulin may bind non-specifically to Fc receptors.
Host Species: Mouse
Clone: MOPC-173
Isotype: IgG2a κ
Flow Cytometry, Intracellular Staining
Specifications
Specifications
| Applications | Flow Cytometry, Western Blot |
| Clone | MOPC-173 |
| Conjugate | Alexa Fluor 647 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.