Learn More
Description
Intracellular Staining, Isotype Control
Specifications
Specifications
| Applications | Western Blot |
| Clone | MOPC-173 |
| Conjugate | Alexa Fluor 700 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | 4°C |
| Isotype | IgG2a |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.