Learn More
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | G155-178 |
| Concentration | 0.2mg/mL |
| Conjugate | Pacific Blue |
| Host Species | Mouse |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2a |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.