Learn More
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This immunoglobulin is specific for trinitrophenol (TNP), a hapten not expressed on human cells or human cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors. This antibody is routinely tested on peripheral blood by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | G155-178 |
| Conjugate | PE-Cyanine7 |
| Host Species | Mouse |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2a |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.