Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2a κ, Mouse, V450, Clone: MOPC-173, BD

Catalog No. BDB560882 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560882 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560882 Supplier BD Biosciences Supplier No. 560882
Add to Cart
Add to Cart

Intracellular Staining, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Western Blot
Clone MOPC-173
Conjugate Violet Fluor 450 (V450)
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Content And Storage 4°C
Isotype IgG2a κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.