Learn More
Description
Flow Cytometry
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | X39 |
| Conjugate | PE |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 25 μg |
| Primary or Secondary | Isotype Control (Primary) |
| Content And Storage | Undiluted at 2° to 8°C. Protect from exposure to light. Do not freeze. |
| Isotype | IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.