Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2a, λ, PE-Cy™7, Clone: B39-4, BD

Catalog No. BDB560721 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560721 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560721 Supplier BD Biosciences Supplier No. 560721
Add to Cart
Add to Cart

The B39-4 antibody is specific for keyhole limpet hemocyanin (KLH).

Control, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a λ
Classification Monoclonal
Clone B39-4
Concentration 0.2mg/mL
Conjugate PE
Host Species Rat
Immunogen Trinitrophenol-KLH
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.