Learn More
anti-IgG2a, κ, R-PE, Clone: G155-178, BD
Routinely tested by flow cytometric analysis. anti-IgG2a, κ, R-PE, Clone: G155-178, BD was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Supplier: BD Biosciences 555574
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Applications: Flow Cytometry, Isotype Control

Specifications
Flow Cytometry | |
R-PE | |
Affinity Purified | |
Isotype Control (Primary) | |
IgG2a κ |
G155-178 | |
Mouse | |
100 Tests | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.