Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2a κ, Rat , Alexa Fluor™ 647, Clone: R35-95, BD

Catalog No. BDB557906 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
100 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557906 100 Tests
BDB557690 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB557906 Supplier BD Biosciences Supplier No. 557906
Add to Cart
Add to Cart

The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κImmunogen: Mouse Pooled Immunoglobulin

Flow Cytometry, Intracellular Staining, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry, Western Blot
Clone R35-95
Conjugate Alexa Fluor 647
Host Species Rat
Immunogen Mouse Pooled Immunoglobulin
Purification Method Affinity Purified
Quantity 100 Tests
Content And Storage Undiluted at 4°C
Isotype IgG2a κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.