Learn More
Description
The B39-4 antibody is specific for keyhole limpet hemocyanin (KLH).
Host Species: Rat
Clone: B39-4
Isotype: IgG2a, <special id='310'/>Immunogen: Trinitrophenol-KLH
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | B39-4 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Rat |
| Immunogen | Trinitrophenol-KLH |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2a |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.