Learn More
anti-IgG2a κ, Rat , PE, Clone: R35-95, BD
Supplier: BD Biosciences 557229
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This reagent has been used as an isotype control in immunophenotypic studies with baboon, rhesus, or cynomolgus peripheral blood cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Isotype Control

Specifications
Flow Cytometry | |
PE | |
Mouse Pooled Immunoglobulin | |
50 Tests | |
Undiluted at 4°C |
R35-95 | |
Rat | |
Affinity Purified | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.