Learn More
Description
The MPC-11 antibody has unknown specificity. The MPC-11 cell line was adapted to continuous culture from the tranplantable Merwin Plasmacytoma-11, which was induced by intraperitoneal implantation of a Millipore diffusion chamber into a BALB/c mouse. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | MPC-11 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2b κ |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.