Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG2b κ, Isotype Control, Clone: R35-3, BD

Catalog No. BDB563233 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563233 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563233 Supplier BD Biosciences Supplier No. 563233
Add to Cart
Add to Cart

Host Species: Rat
Clone:R35-3
IsotypeIgG2b, κ

Intracellular Staining, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone R35-38
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.