Learn More
Description
The mouse IgG2b, κ immunoglobulin isotype control monoclonal antibody 27-35 is specific for the hapten dansyl (5-[dimethylamino] naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing which demonstrated low background staining on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 27-35
Isotype: IgG2b, κ
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry, Immunocytology |
| Clone | G155-178 |
| Conjugate | Brilliant Violet 421 |
| Host Species | Mouse |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Content And Storage | 4°C, protect from prolonged exposure to light, DO NOT FREEZE |
| Isotype | IgG2a κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.