Learn More
Description
The A95-1 antibody has unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten that is not expressed on human, mouse, rat, or non-human primate cells. The A95-1 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues
Host Species: Rat
Clone: A95-1
Isotype: IgG2b κImmunogen: TNP-Keyhole Limpet Hemocyanin
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | A95-1 |
| Concentration | 0.2mg/mL |
| Conjugate | Alexa Fluor 700 |
| Host Species | Rat |
| Immunogen | TNP-keyhole limpet hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG2b |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.