Learn More
Description
Mouse IgG3 κ isotype control antibody is specific for the β-2 - 6-linked Fructosan. The transplantable plasmacytoma J606 was induced by inraperitoneal injection of mineral oils into BALB/c mice. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | J606 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Target Species | Human |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG3 κ |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.