Learn More
Description
The A112-3 clone is specific for keyhole limpet hemocyanin, (KLH). TNP-KLH, the immunogen, is not expressed on human, mouse, or rat cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone A112-3 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody
development only or reported in the literature.
Flow Cytometry, Immunohistochemistry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry, Immunohistochemistry |
| Clone | A112-3 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Content And Storage | Undiluted at 4°C |
| Isotype | IgG3 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.