Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG3 κ, Mouse, Clone: J606, NA/LE, Isotype Control, BD

Catalog No. BDB550341 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550341 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550341 Supplier BD Biosciences Supplier No. 550341
Add to Cart
Add to Cart

Mouse IgG3 κ isotype control antibody is specific for the β-2 - 6-linked Fructosan. The transplantable plasmacytoma J606 was induced by inraperitoneal injection of mineral oils into BALB/c mice. This antibody is routinely tested by immunohistochemistrical analysis on frozen rat tissue sections. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: J606
Isotype: IgG3, κ

Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed), Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Zinc-Fixed)
Clone J606
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 1 mL
Content And Storage Undiluted at 4°C
Isotype IgG3 κ
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.