Learn More
Description
Host Species: Mouse
Clone:G155-228
IsotypeIgM, κ
Immunogen:TNP-Keyhole Limpet Hemocyanin (KLH)
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | G155-228 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 510 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | TNP-Keyhole Limpet Hemocyanin |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.