Learn More
Description
The R4-22 antibody has unknown specificity. This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488nm), green (532nm) and yellow-green (561nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20nm filter).
Host Species: Rat
Clone: R4-22
Isotype: IgM, κ
Immunogen: Pooled Mouse Ig
Flow Cytometry, Isotype Control
Specifications
Specifications
| Antigen | IgM, κ |
| Classification | Monoclonal |
| Clone | R4-22 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-CF594 |
| Host Species | Rat |
| Immunogen | Pooled Mouse Ig |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.