Learn More
anti-IgM κ, Mouse, Clone: G155-228, Isotype Control, BD
Supplier: BD Biosciences 553472
Description
The G155-228 antibody is specific for the hapten trinitrophenol (TNP), a hapten not expressed on non-human primate cells or non-human primate cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors. Th
Host Species: Mouse
Clone: G155-228
Isotype: IgM, κ
Species Reactivity: Mouse
Immunogen: TNP-Keyhole Limpet Hemocyanin
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Intracellular Staining

Specifications
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.5mg/mL | |
Mouse | |
Affinity Purified | |
Mouse | |
IgM κ |
G155-228 | |
Unconjugated | |
TNP-keyhole limpet hemocyanin | |
0.5 mg | |
Undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.