Learn More
Description
The R4-22 antibody has unknown specificity.
Host Species: Rat
Clone: R4-22
Isotype: IgM, κ
Immunogen: Pooled Mouse Ig
Flow Cytometry, Isotype Control
Specifications
Specifications
| Applications | Flow Cytometry |
| Clone | R4-22 |
| Concentration | 0.2mg/mL |
| Conjugate | APC-Cyanine7 |
| Host Species | Rat |
| Immunogen | Pooled Mouse Ig |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Content And Storage | Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
| Isotype | IgM κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.