Learn More
anti-IgM κ, Rat, Clone: R4-22, Isotype Control, BD
Supplier: BD Biosciences 550342
Description
The R4-22 antibody has unknown specificity. This antibody is routinely tested by immunohistochemical analysis on paraffin-embedded tissue sections. Other applications were tested at BD
Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R4-22
Isotype: IgM, κ
Immunogen: Pooled Mouse Ig
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (Zinc-Fixed), Isotype Control, Flow Cytometry

Specifications
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Pooled Mouse Ig | |
1 mL | |
IgM κ |
R4-22 | |
Rat | |
Affinity Purified | |
Undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.