Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1β Mouse anti-Human, Unlabeled, Clone: AS10, BD

Catalog No. BDB550007 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550007 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550007 Supplier BD Biosciences Supplier No. 550007
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AS10 antibody reacts with human interleukin-1β (IL-1β) which is also known as endogenous pyrogen (EP), leukocyte endogenous mediator (LEM), mononuclear cell factor (MCF) and lymphocyte-activating factor (LAF). IL-1β is a proinflammatory cytokine that is synthesized as a precursor of 31kDa and is converted intracellularly to the mature 17.5kDa form, after cleavage by the IL-1β-converting enzyme (ICE). In healthy individuals, IL-1β is secreted non-constitutively by blood monocytes, tissue macrophages and dendritic cells. IL-1β is also constitutively expressed in the human hypothalamus. Many malignant tumors express IL-1β as part of their neoplastic nature. The immunogen used to generate the AS10 hybridoma was recombinant human E. Coli-expressed recombinant IL-1β. The AS10 antibody does not cross react with mouse IL-1β. This is a neutralizing antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1β
Applications Immunocytochemistry
Classification Monoclonal
Clone AS10
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant human IL-1β
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.