Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD HiCK-3 Human Cytokine Positive Control Cells

Catalog No. bdb555063 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555063 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555063 Supplier BD Supplier No. 555063
Add to Cart
Add to Cart
  • This suspension contains Human intracellular CytoKine-3 (HiCK-3) Positive Control Cells
  • The HiCK-3 frozen cell suspension contains fixed, non-permeabilized human lymphoid cells
  • The suspension includes cells that express detectable levels of intracellular IL-1α, IL-1β, IL-6, IL-12 p40/p70, IL-12 p70, and TNF as determined by immunofluorescent intracellular cytokine staining and flow cytometry
  • HiCK-3 cells contain a measurable proportion of cytokines
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 x106 cells/mL
For Use With (Application) Intracellular staining (flow cytometry)
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.