Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-13 Mouse anti-Human, Biotin, Clone: B69-2, BD

Catalog No. BDB555054 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555054 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555054 Supplier BD Biosciences Supplier No. 555054
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B69-2 antibody reacts with human interleukin-13 (IL-13). The immunogen used to generate the B69-2 hybridoma was recombinant human IL-13 protein. This antibody is routinely tested by ELISA Detection. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-13
Applications ELISA
Classification Monoclonal
Clone B69-2
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant human IL-13 protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.