Learn More
AdipoGen anti-IL-1alpha (p18) (mouse), mAb (Teo-1)
SDP
Supplier:  AdipoGen AG20B0064C100
Encompass_Preferred
anti-IL-1alpha (p18) (mouse), mAb (Teo-1), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes mouse IL-1alpha p18 cleaved and full-length fragments. Isotype: Mouse IgG. Clone: Teo-1. Applications: ELISA, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. The most prominent members of the interleukin-1 (IL-1) superfamily are IL-1alpha and IL-1beta. They lack a signal peptide and are secreted by an unconventional, endoplasmic reticulum-Golgi-independent mechanism. IL-1alpha was reported to be more widely and constitutively expressed and has intracellular functions, but also acts locally in a membrane-bound form by activating IL-1R1. Additionally, passive release of IL-1alpha upon cell death can trigger a sterile inflammatory response to dying cells. The cleavage of IL-1alpha is not mediated by caspase-1 and is not required for binding to IL-1R1.
Catalog No. 50-101-5690
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.