Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-21 R, PE, Clone: 155516, R&D Systems™

Catalog No. FAB5961P025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5961P025 25 Tests
FAB5961P100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB5961P025 Supplier R&D Systems Supplier No. FAB5961P025
This item is not returnable. View return policy

Specifications

Antigen IL-21 R
Applications Flow Cytometry
Classification Monoclonal
Clone 155516
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9JHX3
Gene Alias CD360, CD360 antigen, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor
Gene Symbols IL21R
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-21 R, Cys20-Pro236, Accession # Q9JHX3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50615
Test Specificity Detects mouse IL-21 R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human IL-21 R or recombinant mouse IL-2 R beta is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.