Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL1RN Matched Antibody Pair, Ab-1, Abnova

Catalog No. 89017296 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-296 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-296 Supplier Abnova Corporation Supplier No. H00003557AP11
Only null left
Add to Cart
Add to Cart

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-IL1RN (100μg); Mouse monoclonal anti-IL1RN, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This protein inhibits the activities of interleukin 1, α (IL1A) and interleukin 1, β (IL1B), and modulates a variety of interleukin 1 related immune and inflammatory responses. This gene and five other closely related cytokine genes form a gene cluster spanning approximately 400 kb on chromosome 2. A polymorphism of this gene is reported to be associated with increased risk of osteoporotic fractures and gastric cancer. Four alternatively spliced transcript variants encoding distinct isoforms have been reported.

ELISA

Specifications

Antigen IL1RN
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL1RN.
Gene IL1RN
Gene Alias ICIL-1RA/IL-1ra3/IL1F3/IL1RA/IRAP/MGC10430
Gene Symbols IL1RN
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Research Discipline Membrane Receptors
Primary or Secondary Primary
Gene ID (Entrez) 3557
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IL1RN (100 μg) 2. Detection antibody: mouse monoclonal anti-IL1RN, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.