Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILK Mouse, Unlabeled, Clone: 3, BD

Catalog No. BDB611803 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611803 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611803 Supplier BD Biosciences Supplier No. 611803
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrins are transmembrane receptors that mediate cell-cell or cell-matrix adhesion. All integrins are heterodimers composed of α and β subunits, which interact with extracellular matrix and cytoskeletal proteins. Signal transduction through intergin receptors may be regulated by integrin-linked kinase (ILK). ILK is a widely expressed Ser/Thr protein kinase that contains four ankyrin-like repeats in the N-terminal region, a phoshoinositide lipid-binding motif at amino acids 180-212, and an integrin binding site at amino acids 293-451. The ankyrin repeats interact with a LIM domain-only protein called PINCH that also binds the adaptor protein, Nck-2. This interaction implicates ILK in growth factor receptor pathways. In addition, the interaction of ILK with the cytoplasmic domains of integrin β1 and β3 subunits and localization of ILK to focal adhesion plaques suggests a role for ILK in the regulation of cell adhesion. Interestingly, overexpression of ILK in epithelial cells inhibits cell adhesion and stimulates cell cycle progression and tumorigenicity. Thus, ILK regulates signal transduction in many different cell signaling pathways.

Host Species: Mouse
Clone: 3
Isotype: IgG2
Species Reactivity [for Features Main]: Human
Immunogen: Human ILK aa. 326-452

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ILK
Applications Western Blot
Classification Monoclonal
Clone 3
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ILK aa. 326-452
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.