Learn More
Advanced ImmunoChemical, Inc. ANTI-IMMUNE COMPLEX MAB
Supplier:  Advanced ImmunoChemical, Inc. 2IC1 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs:Ab-BNP2ccHybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with immune complex formed by Fab-fragment of BNP-specific MAb 24C5 (Cat.# 2-BNP) and synthetic human BNP.Specificity:Human BNP and proBNP in immune complex with MAb 24C5MAb Isotypes:IgG2a for MAb Ab-BNP2ccApplications:BNP and proBNP sandwich immunoassays (detection), only as a pair to MAb 24C5.Purification:Protein A affinity chromatographyPresentation:PBS, pH 7.4, 0.09 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566221
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.