Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Gloves with Detachable Wrist Support

Catalog No. 19031925 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Large
Hand:
Left
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-031925 Large Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-031925 Supplier Impacto™ Supplier No. 70420120041
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Grain leather on the palm provides abrasion protection, while the detachable wrist support helps prevent repetitive strain injuries.

  • VEP padding in the palm and thumb web area
  • Detachable grain leather wrist support
  • Fingerless style for optimum dexterity
  • Protection from abrasion, impact and shock
  • Minimizes extension and over flexion of the wrist to prevent RSI
  • Use with hand or electric tools in repetitive tasks

Specifications

Back Material Spandex
Cuff Material Grain Leather
Color Blue, Yellow
Hand Left
Material Grain Leather
Product Type Anti-impact Gloves
Size Large
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Color Yellow
Series 704-20
Length (English) 9 in.
Length (Metric) 22.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.