Learn More
Advanced ImmunoChemical, Inc. ANTI-INFLUENZA A HAEM H5
Catalog No. NC3566235 Shop All Advanced ImmunoChemical, Inc. Products
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs:1C7, 1B4. Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified avian influenza virus, type A (H5N1).Specificity:MAbsreactwithhaemagglutininH5.Mouse Isotypes:1C7, 1B4 – IgG2aApplications:MAbs 1C7 can be used in haemagglutinin inhibition (HI) test. MAb 1B4 can be used in ELISA.Recommended pair for sandwich immunoassay (capture – detection): 1B4 – 1C7.Purification:Protein G chromatography.Presentation:PBS, pH 7.4, 0.09% sodium azide (NaN3).MAb: Concentration: mg/mlLot: Volume: mlExpiration date: Amount: mgStorage Conditions:Store @ +4oC (+2 ̊…+8 ̊C allowable).
Catalog No. NC3566235 Supplier Advanced ImmunoChemical, Inc. Supplier No. 5IVAH51 MG
MONOCLONAL ANTIBODIES/MAbs:1C7, 1B4. Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified avian influenza virus, type A (H5N1).Specificity:MAbsreactwithhaemagglutininH5.Mouse Isotypes:1C7, 1B4 – IgG2aApplications:MAbs 1C7 can be used in haemagglutinin inhibition (HI) test. MAb 1B4 can be used in ELISA.Recommended pair for sandwich immunoassay (capture – detection): 1B4 – 1C7.Purification:Protein G chromatography.Presentation:PBS, pH 7.4, 0.09% sodium azide (NaN3).MAb: Concentration: mg/mlLot: Volume: mlExpiration date: Amount: mgStorage Conditions:Store @ +4oC (+2 ̊…+8 ̊C allowable).