Learn More
Advanced ImmunoChemical, Inc. ANTI-INFLUENZA B MAB
Supplier: Advanced ImmunoChemical, Inc. 5IBV1 MG
MONOCLONAL ANTIBODIES/MAbs:R2/3Specificity:MAbs react with hemaglutinin H5 in hemagluttinin inhibition (HI) test.Applications:Indirect EIA, Western blottingPurification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.