Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD11b/Integrin alpha M Fluorescein-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4715624
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1124F025 25 Tests
FAB1124F100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1124F025 Supplier R&D Systems Supplier No. FAB1124F025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

CD11b Monoclonal specifically detects CD11b in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD11b
Applications Flow Cytometry
Classification Monoclonal
Clone M1/70
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Gene Symbols ITGAM
Host Species Rat
Immunogen Con A-activated C57BL/10 splenocytes
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3684
Test Specificity Detects mouse Integrin alpha M/CD11b. Cross-reaction with human Integrin alpha M has been reported (1, 2).
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.