Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin αVΒ3 Mouse anti-Human, FITC, Clone: LM609, MilliporeSigma™

Catalog No. FCMAB282FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB282FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB282FMI Supplier MilliporeSigma Supplier No. FCMAB282F
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Integrin αVβ3 using this Milli-Mark™ Anti-Integrin αVβ3 Antibody, clone LM609-FITC validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin αVβ3
Applications Flow Cytometry
Classification Monoclonal
Clone LM609
Conjugate FITC
Formulation Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_002201
Gene Symbols ITGAV, DKFZp686A08142, MSK8, CD51, VNRA
Host Species Mouse
Immunogen Purified adhesion receptor from the human melanoma cell line M21 (Cherish, D. and Spiro, R. (1987) J. Biol. Chemistry, 262(36):17703-17711).
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.