Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP3R-3 Mouse, Unlabeled, Clone: 2, BD

Catalog No. BDB610312 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610312 50 μg
BDB610313 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610312 Supplier BD Biosciences Supplier No. 610312
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Inositol 1,4,5-triphosphate (IP3) functions as a second messenger for many hormones, growth factors, and neurotransmitters. IP3 causes the release of Ca2+ from intracellular stores by binding specific receptors that are coupled to Ca2+ channels. A number of studies have identified a family of at least four IP3 receptors (IP3R). The type III receptor (IP3R-3) has been isolated and characterized in human and rat. IP3 receptors are commonly localized in the endoplasmic reticulum, but have also been identified in the nucleus and the plasma membrane. Co-expression of different IP3 receptors is detected in most tissues and cell lines. Although these receptors appear to have a similar specificity for inositol phosphates, the different receptors have been reported to have different affinities for IP3 as follows: type II > type I > type III.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IP3R-3
Applications Western Blot
Classification Monoclonal
Clone 2
Concentration 250μg/mL
Conjugate Unconjugated
Description Inositol 1,4,5-triphosphate receptor type III
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.