Learn More
Description
- Anti-Ipilimumab (Yervoy™) (Human) ELISA Kit is a sandwich enzyme-linked immunoassay for the qualitative measurement of antibody against Ipilimumab in human serum or plasma.
Specifications
Specifications
| Cross Reactivity | Human |
| Product Type | Kit |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.