Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isocitrate Dehydrogenase 1 (IDH1)-R132H Mouse anti-Human, Clone: HMab-1, MilliporeSigma™

Catalog No. MABC171MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC171MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC171MI Supplier MilliporeSigma Supplier No. MABC171
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Isocitrate Dehydrogenase 1 (IDH1)-R132H using this Anti-Isocitrate Dehydrogenase 1 (IDH1)-R132H Antibody, clone HMab-1 validated for use in Western Blotting, immunohistochemistry, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Isocitrate Dehydrogenase 1 (IDH1)-R132H
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone HMab-1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005887
Gene Symbols IDH1, PICD
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Isocitrate Dehydrogenase 1 (IDH1).
Purification Method Protein G purified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.