Learn More
Diagnostic Biosystems ANTI ISOCITRATE DEHYDROGENENAS Anti-Isocitrate Dehydrogenenase 1 (IDH1)-R132S
Supplier:  Diagnostic Biosystems MOB582R01
Encompass_Preferred
Isocitrate dehydrogenase 1 (IDH1) is a 46 kDa NADP-dependent enzyme, which catalyzes the decarboxylation of isocitrate into α-ketoglutarate. IDH1 may also play a role in the prevention of oxidative damage, and the shuttling of proteins to peroxisomes. It is widely reported that mutations in IDH1 result in multiple forms of gliomas. The Arginine to Serine substitution at amino acid 132 is seen in gliomas, abolishes magnesium binding and alters enzyme activity so that isocitrate is no longer converted to alphaketoglutarate but instead alpha-ketoglutarate is converted to R(-)-2- hydroxyglutarate.
Catalog No. 50-238-5465
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.