Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAM-1 Mouse anti-Human, Unlabeled, Clone: 43, BD

Catalog No. BDB612120 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612120 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612120 Supplier BD Biosciences Supplier No. 612120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tight junctions (zonulae occludens) are critical to the maintenance of cell polarity and intercellular barriers. Protein components of the tight junctions include actin filaments, symplekin, occludin, ZO-1, ZO-2, and ZO-3. Junctional adhesion molecules, JAM-1, -2, and -3, are a new subfamily in the immunoglobulin (Ig) superfamily that colocalize with these components of tight junctions. JAM-1 was identified through a screen of antibodies that recognizes antigens located at cell-cell contacts. The structure of JAM-1 includes two V-type Ig domains in the extracellular region, two N-glycosylation sites, and a small C-terminal cytoplasmic region. JAM-1 protein is expressed at cell-cell junctions in endothelial and epithelial cells, and antibodies to JAM-1 inhibit spontaneous and chemokine-induced monocyte transmigration through an endothelial cell monolayer. In addition, TNFα and IFN-γ cause redistribution of JAM-1 from intercellular junctions to the cytoplasm. JAM-1 also is the receptor for the platelet membrane protein, F11 antigen, and activation of platelets leads to PKC phosphorylation of JAM-1. Thus, JAM-1 is a junctional adhesion molecule that functions in endothelial, epithelial, and platelet adhesion.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen JAM-1
Applications Western Blot
Classification Monoclonal
Clone 43
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human JAM aa. 126-237
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.