Learn More
Selleck Chemical LLC Anti-KAAG1-D4062-1MG-5
SDP
Supplier:  Selleck Chemical LLC D40621MG5
Encompass_Preferred
Anti-KAAG1 is an ADC composed of a humanized monoclonal antibody (3A4) targeting human KAAG1 It is used for the treatment of advanced solid tumors including platinum resistant ovarian cancer and triple negative breast cancer MW 145 28 KD
Catalog No. 50-313-4611