Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

katanin p60 (ATPase-containing) subunit A 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017018 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-018 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-018 Supplier Abnova Corporation Supplier No. H00011104B01P
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human KATNA1 protein.

Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60kDa ATPase (p60 subunit A 1) and an 80kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases. [provided by RefSeq

Sequence: MSLLMISENVKLAREYALLGNYDSAMVYYQGVLDQMNKYLYSVKDTYLQQKWQQVWQEINVEAKHVKDIMKTLESFKLDSTPLKAAQHDLPASEGEVWSMPVPVERRPSPGPRKRQSSQYSDPKSHGNRPSTTVRVHRSSAQNVHNDRGKAVRCREKKEQNKGREEKGVLMVGPPGTGKTLLAKAVATECKTTFFNVSSSTLTSKYRGESEKLVRLLFEMARFYSPATIFIDEIDSICSRRGTSEEHEASRRVKAELLVQMDGVGGTSENDDPSKMVMVLAATNFPWDIDEALRRRLEKRIYIPLPSGMRP

Specifications

Antigen katanin p60 (ATPase-containing) subunit A 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human KATNA1 protein.
Formulation PBS with no preservative; pH 7.4
Gene KATNA1
Gene Accession No. BC050428.1
Gene Symbols KATNA1
Host Species Mouse
Immunogen KATNA1 (AAH50428.1, 1 a.a. ∼ 311 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 11104
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.