Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KCTD4, Human, IP-WB Antibody Pair, Abnova™

Numéro de catalogue. 89301744 missing translation for 'brandsLinkText'
Modifier l'affichage
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
89-301-744 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 89-301-744 Fournisseur Abnova Corporation Code fournisseur H00386618PW2
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Comprend

Antibody pair for IP: rabbit polyclonal anti-KCTD4 (300μL), Antibody pair for WB: mouse purified polyclonal anti-KCTD4 (50μg)

For immunoprecipitation and to detect the precipitated protein in western blot.

Spécifications

Antigen KCTD4
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene KCTD4
Gene Alias bA321C24.3
Gene Symbols KCTD4
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 386618
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-KCTD4 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-KCTD4 (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Afficher plus de résultats Afficher moins de résultats

For Research Use Only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.