Learn More
Description
Specifications
Specifications
| Antigen | Ki-67 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 10mM PBS, pH 7.4 containing BSA and sodium azide. |
| Gene Symbols | MKI67; KIA; Ki-67 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from human Ki-67 |
| Purification Method | Affinity Purified |
| Quantity | 500 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.