Learn More
Advanced ImmunoChemical, Inc. ANTI-KIDNEY INJURY MOLECULE
Catalog No. NC3566224 Shop All Advanced ImmunoChemical, Inc. Products
Encompass_Preferred
MONOCLONAL ANTIBODIES/MAbs:KIM70, KIM75Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human recombinant KIM-1 ectodomain expressed in baculovirus system.Specificity:Human KIM-1MAb Isotypes:IgG1 for MAbs KIM70, KIM75Applications:MAbs KIM70, KIM75 react with human recombinant KIM-1 in sandwich ELISA and Western blotting.Recommended pairs KIM-1 immunodetection in sandwich immunoassay (capture-detection):KIM70 – KIM75Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C
Catalog No. NC3566224 Supplier Advanced ImmunoChemical, Inc. Supplier No. 2KIM11 MG
MONOCLONAL ANTIBODIES/MAbs:KIM70, KIM75Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human recombinant KIM-1 ectodomain expressed in baculovirus system.Specificity:Human KIM-1MAb Isotypes:IgG1 for MAbs KIM70, KIM75Applications:MAbs KIM70, KIM75 react with human recombinant KIM-1 in sandwich ELISA and Western blotting.Recommended pairs KIM-1 immunodetection in sandwich immunoassay (capture-detection):KIM70 – KIM75Purification:Chromatography on protein A SepharosePresentation:PBS, pH 7.4, 0.1 % sodium azide (NaN3)Storage Conditions:+ 4 °C