Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-KLH, PE-CF594, Clone: X40, BD

Catalog No. BDB562292 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562292 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562292 Supplier BD Biosciences Supplier No. 562292
Add to Cart
Add to Cart

Host Species: Mouse
Clone: X40
Isotype: IgG1, κ
Immunogen: Keyhole limpet hemocyanin

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Antigen KLH
Classification Monoclonal
Clone X40
Concentration 0.2mg/mL
Conjugate PE-CF594
Host Species Mouse
Immunogen Keyhole limpet hemocyanin
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.