Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda, Ig Light Chain Mouse anti-Human, PE, Clone: 1-155-2, BD

Catalog No. BDB642919 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB642919 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB642919 Supplier BD Biosciences Supplier No. 642919
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Lambda is specific for lambda light chains of human immunoglobulins. Immunoglobulins bearing lambda light chains are present on approximately 40% of normal B lymphocytes and on Igλ+ neoplastic cells. In serum, Anti-Lambda reacts with immunoglobulins bearing lambda light chains as well as free lambda light chains.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lambda, Ig Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone 1-155-2
Concentration 12.5μg/mL
Conjugate PE
Formulation Phosphate buffered saline with BSA and 0.1% sodium azide.
Host Species Mouse
Immunogen Human IgA
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 λ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.