Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Β1 Mouse anti-Human, Clone: 4E10, MilliporeSigma™

Catalog No. MAB1921PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1921PMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1921PMI Supplier MilliporeSigma Supplier No. MAB1921P
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Laminin β1 using this Anti-Laminin β1 Antibody, clone 4E10 validated for use in ELISA, immunofluoresence, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin β1
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4E10
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS, pH 7.6, containing 0.1% sodium azide.
Gene Accession No. NM_002291.1
Gene Symbols LAMB1, MGC142015, CLM
Host Species Mouse
Immunogen Purified human laminin
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.