Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin α2 Mouse anti-Human, Monkey, Rabbit, Clone: 5H2, MilliporeSigma™

Catalog No. MAB1922ZMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1922ZMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1922ZMI Supplier MilliporeSigma Supplier No. MAB1922Z
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Laminin α2 Antibody, clone 5H2, Azide Free detects level of Laminin α2 and has been published and validated for use in ELISA, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin α2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5H2
Conjugate Unconjugated
Gene Accession No. NM_000426.3; NM_001079823.1
Gene Symbols LAMA2, LAMM
Host Species Mouse
Immunogen Purified human merosin.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Monkey, Rabbit
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.